Potassium iodide ≥99%, crystals ACS
Supplier: Spectrum Chemical Mfg. Corp.
Potassium Iodide, Crystal, Reagent, ACS, is reffered to by the chemical formula KI and is an inorganic compound. Naturally occuring in Kelp, this white salt is one of the most significant iodide compounds. Produced by treating KOH with iodine, this compound has a variety of uses. Used as a component in film photography before the discovery of silver iodide, it is also used in the electrolyte of dye sensitized solar cells. KI is also used as an emergency blockade of radio-iodine, a common form of radiation poisoning. As an ACS grade Reagent, Spectrum Chemical manufactured Potassium Iodide is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness
Formula:
Kl Boiling Pt: 1330 °C (1013 hPa) Melting Pt: 686 °C Density: 3.13 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011405 CAS Number: 7681-11-0 EINECS: 231-659-4 Merck Index: 13,07727 |
Specification Test Results
Assay | 99% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...