Potassium hydroxide ≥85%, pellets ACS suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

P1315-2.5KG P1315-125GM P1315-500GM P1315-12KG P1315-1KG
700000-766EA 357.24 USD
700000-766 700000-760 700000-768 700000-762 700000-764
Potassium hydroxide ≥85%, pellets ACS suitable for use as excipient
Potassium hydroxide

Potassium Hydroxide, Pellets, Reagent, ACS is an inorganic compound commonly referred to as Caustic Potash. A colorless solid chemical compound, POTASSIUM HYDROXIDE, PELLETS, Reagent, ACS is a protypical strong base. In the case of POTASSIUM HYDROXIDE, PELLETS, Reagent, ACS it's commonly used as a precursor to soft and liquid soaps, as well as potassium containing chemicals. ACS or Reagent grade Potassium Hydroxide is used in the laboratory as an analytical testing reagent

Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Density: 2.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8 II
REACH: 01-2119487136-33
Merck Index: 12,07806

Order Now

Specification Test Results

Assay 85% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR