2-Pentanone ≥90% for synthesis
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
Synonyms:
Methyl Propyl Ketone
P1019-200LTB
P1019-20LTBL
700003-946EA
3798.9
USD
700003-946
700003-948
2-Pentanone ≥90% for synthesis
2-Pentanone
2-Pentanone is chemically a ketone and considered to be similar in type to methylethylketone but is more expensive and has a lower solvency. It is found in leaf tobacco. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Formula:
C₅H₁₀O Boiling Pt: 100 °C (1013 hPa) Melting Pt: –78 °C Density: 0.809 g/cm³ (20 °C) Flash Pt: 7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009400 CAS Number: 107-87-9 EINECS: 203-528-1 UN: 1249 ADR: 3,II Merck Index: 12,06193 |
Specification Test Results
Assay | 90% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...