Nitric acid 70% ≥70% ACS
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
Synonyms:
Nitric acid solution
N1075-2.5LTP
N1075-500MLG
700000-594EA
253.98
USD
700000-594
700000-596
700000-600
Nitric acid 70% ≥70% ACS
Nitric acid
Nitric Acid, 70 Percent, Reagent, ACS is a strong and highly corrosive mineral acid. Primarily used as a reagent, the pure compound is colorless and used for nitration, typically to an organic molecule. It is also used as a strong oxidizing agent due to its corrosive properties. As an ACS grade Reagent, Spectrum Chemical manufactured Nitric Acid is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and purity
Formula:
HNO₃ MW: 63.01 g/mol Density: 1.400 Storage Temperature: Ambient |
MDL Number:
MFCD00011349 CAS Number: 7697-37-2 EINECS: 231-714-2 UN: 2031 ADR: 85.1,II |
Specification Test Results
Assay | 68% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...