2-Methylbutane ≥98%, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: iso-Pentane

M1246-200LTB M1246-20LT M1246-500ML M1246-4LTGL M1246-100ML
700002-204EA 2876.55 USD
700002-204 700002-206 700002-210 700002-208 700002-202
2-Methylbutane ≥98%, Reagent Grade
2-Methylbutane

2-Methylbutane, Reagent is also known under the name Isopentane and is an extremely volatile and flammable liquid. It is used with liquid nitrogen to obtain liquid bath temperatures of -160ºC and in the production of dental products Aquafresh and Sensodyne

Formula: CH₃CH₂CH(CH₃)₂
MW: 72.15 g/mol
Boiling Pt: 28 °C (1013 hPa)
Melting Pt: –160 °C
Density: 0.620
Flash Pt: –51 °C
Storage Temperature: Ambient
MDL Number: MFCD00009338
CAS Number: 78-78-4
EINECS: 201-142-8
UN: 1265
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=498904a9f8b2618e19d2343aa74e247ec7121f00&vid=c7ed2044255d0d72af23a3fafec4ef6066d25097&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR