Isoamyl acetate ≥99%, Reagent Grade
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
Synonyms:
Isopentyl acetate, Acetic acid isoamyl ester
Isoamyl Acetate, Reagent, also known as isopentyl acetate, is the ester formed from acetic acid and isoamyl alcohol. Isoamyl acetate is used to confer banana flavor in foods and as a commercial solvent. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
Formula:
C₇H₁₄O₂ MW: 130.19 g/mol Boiling Pt: 142 °C (1013 hPa) Melting Pt: –78 °C Density: 0.876 Flash Pt: 25 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008946 CAS Number: 123-92-2 EINECS: 204-662-3 UN: 1104 ADR: 3,III Merck Index: 12,05125 |
Specification Test Results
Assay | 99% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...