(±)-2-Methyl-2,4-pentanediol ≥98%, Reagent Grade suitable for use as excipient
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
Hexylene glycol, (±)-2,4-Dihydroxy-2-methyl pentane, (±)-2,4-Dihydroxy-2-methyl pentane, 2-Methylpentane-2,4-diol, Hexylene Glycol
HE188-20LT
HE188-4LT
HE188-500MLG
HE188-200LTB
700002-872EA
1179.9
USD
700002-872
700002-874
700002-876
700002-870
(±)-2-Methyl-2,4-pentanediol ≥98%, Reagent Grade suitable for use as excipient
(±)-2-Methyl-2,4-pentanediol
Hexylene Glycol, Reagent
Formula:
C₆H₁₄O₂ MW: 118.18 g/mol Boiling Pt: 198 Melting Pt: -40 Density: 0.923 g/cm³ (20 °C) Flash Pt: 93 Storage Temperature: Ambient |
MDL Number:
MFCD00004547 CAS Number: 107-41-5 EINECS: 203-489-0 Merck Index: 13,04730 |
Specification Test Results
Assay | 98% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...