Guanidinium hydrochloride ≥99%, BiotechGrade suitable for use as excipient, for biochemistry

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride

GU103-100GM GU103-2.5KG GU103-500GM GU103-50KGBL
700007-936EA 141.84 USD
700007-936 700007-940 700007-942 700007-944
Guanidinium hydrochloride ≥99%, BiotechGrade suitable for use as excipient, for biochemistry
Guanidinium hydrochloride

Guanidine Hydrochloride, BiotechGrade is considered a chaotrope and a strong denaturants utilized in physiochemical studies of protein folding. Proteins become randomly coiled at high concentrations of guanidinium chloride. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Formula: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Melting Pt: 185
Density: 1.354 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR