D-(+)-Galactose, anhydrous ≥99%, Reagent Grade suitable for use as excipient, for biochemistry

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal

GA110-500GM GA110-100GM GA110-1KG
700009-652EA 536.4 USD
700009-652 700009-648 700009-650
D-(+)-Galactose, anhydrous ≥99%, Reagent Grade suitable for use as excipient, for biochemistry
D(+)-Galactose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 527
Melting Pt: 163…169 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151230
CAS Number: 59-23-4
EINECS: 200-416-4
Merck Index: 13,04356
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=508b1d21a6801ddfb1a5f13a64cb06e6f95e8922&vid=3c02003b1d12c9bfd11c2e6faf5cc37d4325d208&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR