Glyoxal 40% in water

Supplier: Spectrum Chemical Mfg. Corp.
Warning

G1022-3KG G1022-100GM G1022-22KG G1022-500GM G1022-200LTB
700003-708EA 345.95 USD
700003-708 700003-702 700003-706 700003-710 700003-704
Glyoxal 40% in water
Glyoxal

Glyoxal, 40 Percent in Water is used as a crosslinker for starch-based products in the coated paper and textile industry, as well as, being used as a solubilizer and crosslinker in polymer chemistry

Formula: C₂H₂O₂
Density: 1.265
Storage Temperature: Ambient
MDL Number: MFCD00006957
CAS Number: 107-22-2
EINECS: 203-474-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR