Ethylvanillin ≥98% NF suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 3-Ethoxy-4-hydroxybenzaldehyde, 3-(Ethoxy-d5)-4-hydroxybenzaldehyde

ET119-2.5KG ET119-500GM ET119-12KGBL ET119-125GM ET119-40KGBL
95033-162EA 1369.75 USD
95033-162 95033-164 89078-794 95033-160 89078-796
Ethylvanillin ≥98% NF suitable for use as excipient
Ethylvanillin

Ethyl Vanillin, NF is used as a flavorant.


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 285
Melting Pt: 75…77 °C
Flash Pt: 100
Storage Temperature: Ambient
MDL Number: MFCD00006944
CAS Number: 121-32-4
EINECS: 204-464-7
Merck Index: 13,03890

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR