Esculin hydrate for biochemistry

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Aesculin, 6,7-Dihydroxycoumarin 6-glucoside hydrate, Esculin

ES108-100GM ES108-25GM ES108-5GM
700009-494EA 740.8 USD
700009-494 700009-496 700009-498
Esculin hydrate for biochemistry
Esculin hydrate

Esculin is also known as aesculin and occurs naturally in the horse chestnut. It is used in a microbiology laboratory to aid in the identification of bacterial species Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₅H₁₆O₉·xH₂O
Melting Pt: 212 °C
Storage Temperature: Ambient
MDL Number: MFCD00149492
CAS Number: 531-75-9
EINECS: 208-517-5
Merck Index: 12,03739

Order Now

Specification Test Results

MOISTURE (KF) 10.0 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR