Capric acid ≥99%

Supplier: Spectrum Chemical Mfg. Corp.
Warning

D2659-200LTB D2659-20LTBL
700005-104EA 84.73 USD
700005-104 700005-106 700005-108 700005-110
Capric acid ≥99%
Capric acid

Decanoic Acid is also known as capric acid. It is a saturated fatty acid from coconut oil and palm kernel oil. It can also be found in the milk of several types of mammals.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₀H₂₀O₂
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 29…32 °C
Density: 0.893
Flash Pt: 147 °C
Storage Temperature: Ambient
MDL Number: MFCD00004441
CAS Number: 334-48-5
EINECS: 206-376-4
Merck Index: 13,01764

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR