Diethanolamine ≥98.5% ACS suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: ß,ß′-Dihydroxydiethylamine, Bis-(ß-hydroxyethyl)-amine, 2,2'-Iminodiethanol

D1033-4LTGL D1033-100ML D1033-200LTB D1033-500MLG D1033-20LTMT
700006-944EA 1019.5 USD
700006-944 700006-938 700006-940 700006-946 700006-942
Diethanolamine ≥98.5% ACS suitable for use as excipient
Diethanolamine

Diethanolamine, Reagent, ACS is also referred to as DEA, is an organic compound that in its pure form is a white solid but it is most often encountered as a clear liquid that has a viscous texture. It is a secondary amine and diol and is water soluble. It is used as a corrosion inhibitor and surfactant that is used in the oil refining industry to help remove hydrogen sulfide from sour gas. As an ACS grade reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness

Formula: C₄H₁₁NO₂
MW: 105.14 g/mol
Boiling Pt: 268.8
Melting Pt: 28
Density: 1.088
Flash Pt: 176
Storage Temperature: Ambient
MDL Number: MFCD00002843
CAS Number: 111-42-2
EINECS: 203-868-0
REACH: 01-2119488930-28
Merck Index: 13,03134

Order Now

Specification Test Results

Assay 98.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR