Chromium(III) chloride hexahydrate ≥98%, Reagent Grade

Supplier: Spectrum Chemical Mfg. Corp.
Warning

C1238-125GM C1238-12KG C1238-2.5KG C1238-45KGBL C1238-500GM C1238-25GM
700001-126EA 267.93 USD
700001-126 700001-128 700001-130 700001-134 700001-136 700001-132
Chromium(III) chloride hexahydrate ≥98%, Reagent Grade
Chromium (III) chloride hexahydrate

Chromic Chloride, Reagent, is a violet anhydrous compound that can be used as an organic synthesis catalyst and a dye for wool. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Formula: CrCl₃·6H₂O
Melting Pt: 85 °C
Density: 1.76 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149660
CAS Number: 10060-12-5
EINECS: 233-038-3
UN: 3260
ADR: 8,III
Merck Index: 13,02242

Order Now

Specification Test Results

ASSAY (CrCl3•6H2O) 98.0-101.0 %
INSOLUBLE MATTER max. 0.01 %
SUBSTANCES NOT Pptd BY NH4OH (as SO4) max. 0.20 %
SULFATE max. 0.01 %
IRON (Fe) max. 0.01 %
ELEMENTAL IMPURITIES AS REPORTED
IDENTIFICATION TO PASS TEST
RESIDUAL SOLVENTS TO PASS TEST


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR