tert-Butanol ≥99%, Reagent Grade suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: Trimethylcarbinol, tert-Butyl alcohol, 2-Methyl-2-propanol

B1188-20LT B1188-200LTB
700002-686EA 1100 USD
700002-686 700002-684
tert-Butanol ≥99%, Reagent Grade suitable for use as excipient
tert-Butanol

tert-Butyl Alcohol, Reagent, also known as 2-methyl-2-propanol, is a tertiary alcohol that is miscible with diethyl ether and ethanol. It is used as a solvent, ethanol denaturant and paint remover ingredient. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: (CH₃)₃COH
MW: 74.12 g/mol
Boiling Pt: 82.2
Melting Pt: 25.3
Density: 0.775
Flash Pt: 11
Storage Temperature: Ambient
MDL Number: MFCD00004464
CAS Number: 75-65-0
EINECS: 200-889-7
UN: 1120
ADR: 3 II
Merck Index: 12,01577
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63a1b0c51eec643e5d5b7152a3e410db93d1626e&vid=999328a3c31895b190d09313b24501682921af01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR