Benzophenone ≥99%, Reagent Grade for synthesis
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
Diphenylmethanone, Diphenyl ketone
B1088-250GM
B1088-1KG
B1088-25KGBL
700004-026EA
150.65
USD
700004-026
700004-024
700004-028
Benzophenone ≥99%, Reagent Grade for synthesis
Benzophenone
Benzophenone, Reagent, also known as sulisobenzone, is typically an ingredient in most sunscreens. It is used as a skin protectant from damage by short-wave UVA ultraviolet light and UVB. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
Formula:
C₁₃H₁₀O Boiling Pt: 305.4 °C (1013 hPa) Melting Pt: 47…49 °C Density: 1.085 g/cm³ (20 °C) Flash Pt: 150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003076 CAS Number: 119-61-9 EINECS: 204-337-6 UN: 3077 ADR: 9,III Merck Index: 13,01099 |
Specification Test Results
Assay | 99% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...