Bismuth ≥99.5%, powder

Supplier: STREM CHEMICALS INC MS
Warning

93-8345-100G 93-8345-500G
100196-844EA 65.11 USD
100196-844 100198-472
Bismuth ≥99.5%, powder
Bismuth
Formula: Bi
Boiling Pt: 1560 °C
Melting Pt: 271.3 °C
Density: 9,80
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
UN: 3089
ADR: 4,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR