Bismuth(III) oxide ≥99.9% (Bi basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Bismuth Trioxide, Dibismuth trioxide
93-8318-100G
93-8318-500G
100205-798EA
82.09
USD
100205-798
100198-060
Bismuth(III) oxide ≥99.9% (Bi basis)
Bismuth (III) oxide
Formula:
BiO₃ MW: 465.96 g/mol Boiling Pt: 1890 Melting Pt: 825 Density: 8.9 |
MDL Number:
MFCD00003462 CAS Number: 1304-76-3 EINECS: 215-134-7 Merck Index: 13,01273 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...