Bismuth(III) iodide ≥99.999% (Bi basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Bismuth triiodide

93-8317-5G 93-8317-25G
100197-974EA 133.05 USD
100197-974 100209-464
Bismuth(III) iodide ≥99.999% (Bi basis), PURATREM
Bismuth (III) iodide
Formula: BiI₃
Boiling Pt: ~500 °C
Melting Pt: 408 °C
Density: 5.778
MDL Number: MFCD00010894
CAS Number: 7787-64-6
EINECS: 232-127-4
UN: 1759
ADR: 8,III
Merck Index: 12,01308

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR