Lead(II) nitrate ≥99.999% (Pb basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Lead dinitrate
93-8267-25G
93-8267-100G
100208-242EA
147.2
USD
100208-242
100200-352
Lead(II) nitrate ≥99.999% (Pb basis), PURATREM
Lead (II) nitrate
Formula:
Pb(NO₃)₂ MW: 331.21 g/mol Melting Pt: 470 °C (dec.) Density: 4.53 Storage Temperature: Ambient |
MDL Number:
MFCD00011153 CAS Number: 10099-74-8 EINECS: 233-245-9 UN: 1469 ADR: 5.1,II Merck Index: 13,05432 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...