Lead(II) fluoride ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Lead difluoride

93-8214-500G 93-8214-100G
100203-598EA 370.83 USD
100203-598 100205-790
Lead(II) fluoride ≥99%
Lead (II) fluoride
Formula: F₂Pb
MW: 245.2 g/mol
Boiling Pt: 1292
Melting Pt: 824
Density: 8.24
MDL Number: MFCD00011162
CAS Number: 7783-46-2
EINECS: 231-998-8
UN: 2291
ADR: 6.1 III
Merck Index: 12,05425

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR