Gold ≥99.95%, shot

Supplier: STREM CHEMICALS INC MS

93-7913-500MG 93-7913-2G
100210-562EA 157.11 USD
100210-562 100199-192
Gold ≥99.95%, shot
Gold
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR