Gold ≥99.999%, powder
Supplier: STREM CHEMICALS INC MS
93-7902-500MG
93-7902-2G
100205-772EA
220.8
USD
100205-772
100200-808
Gold ≥99.999%, powder
Gold
Formula:
Au MW: 196.97 g/mol Boiling Pt: 2800 Melting Pt: 1063 Density: 19.3 Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 Merck Index: 12,04526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...