Tungsten(IV) carbide ≥99.5% (W basis)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Tungsten monocarbide, Wolfram carbide, Tungsten carbide
93-7431-50G
93-7431-250G
100204-424EA
55.2
USD
100204-424
100205-760
Tungsten(IV) carbide ≥99.5% (W basis)
Tungsten(IV) carbide
Formula:
CW Boiling Pt: 6000 °C Melting Pt: ~2870 °C Density: 15.63 |
MDL Number:
MFCD00011464 CAS Number: 12070-12-1 EINECS: 235-123-0 UN: 3178 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...