Hafnium(IV) bromide, anhydrous ≥98% (99.7%-Hf)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-7217-5G
93-7217-1G
100202-282EA
396.31
USD
100202-282
100204-422
Hafnium(IV) bromide, anhydrous ≥98% (99.7%-Hf)
Hafnium (IV) bromide
Formula:
Br₄Hf Melting Pt: 220 °C (subl.) |
MDL Number:
MFCD00049460 CAS Number: 13777-22-5 EINECS: 237-416-9 UN: 1759 ADR: 8,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...