Erbium triiodide, anhydrous ≥99.9% (Er basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-6823-1G
93-6823-5G
100199-656EA
188.25
USD
100199-656
100207-492
Erbium triiodide, anhydrous ≥99.9% (Er basis)
Erbium triiodide
Formula:
ErI₃ MW: 547.97 g/mol Boiling Pt: 1280 Melting Pt: 1020 °C Density: ∼5.5 g/cm³ (20 °C) |
MDL Number:
MFCD00016075 CAS Number: 13813-42-8 EINECS: 237-471-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...