Holmium ≥99.9%, foil
Supplier: STREM CHEMICALS INC MS
93-6733-25X25MM
93-6733-50X50MM
100199-298EA
348.18
USD
100199-298
100202-716
Holmium ≥99.9%, foil
Holmium
Formula:
Ho Boiling Pt: 2695 °C Melting Pt: 1474 °C Density: 8,795 |
MDL Number:
MFCD00011049 CAS Number: 7440-60-0 EINECS: 231-169-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...