Dysprosium(III) oxide ≥99.998% (dye basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Didysprosium trioxide, Dysprosium oxide
93-6616-25G
93-6616-5G
100199-296EA
403.38
USD
100199-296
100209-190
Dysprosium(III) oxide ≥99.998% (dye basis), PURATREM
Dysprosium (III) oxide
Formula:
Dy₂O₃ MW: 373 g/mol Melting Pt: 2400 Density: 7.81 |
MDL Number:
MFCD00010986 CAS Number: 1308-87-8 EINECS: 215-164-0 Merck Index: 14,03482 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...