Dysprosium(III) oxide ≥99.9% (dye basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Didysprosium trioxide, Dysprosium oxide

93-6615-100G 93-6615-25G
100210-524EA 309.97 USD
100210-524 100205-732
Dysprosium(III) oxide ≥99.9% (dye basis)
Dysprosium (III) oxide
Formula: Dy₂O₃
MW: 373 g/mol
Melting Pt: 2400
Density: 7.81
MDL Number: MFCD00010986
CAS Number: 1308-87-8
EINECS: 215-164-0
Merck Index: 14,03482

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR