Terbium(III) chloride, anhydrous ≥99.9% (Tb basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-6507-10G 93-6507-2G
100208-220EA 561.91 USD
100208-220 100202-252
Terbium(III) chloride, anhydrous ≥99.9% (Tb basis)
Terbium (III) chloride
Formula: Cl₃Tb
Melting Pt: 588 °C
Density: 4.35 g/cm³ (25 °C)
MDL Number: MFCD00011258
CAS Number: 10042-88-3
EINECS: 233-132-4
Merck Index: 13,09087

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR