Terbium(III,IV) oxide ≥99.9% (Tb basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Tetraterbium heptaoxide, Terbium oxide
93-6505-25G
93-6505-5G
100209-182EA
584.55
USD
100209-182
100209-184
Terbium(III,IV) oxide ≥99.9% (Tb basis)
Terbium (III,IV) oxide
Formula:
O₇Tb₄ Melting Pt: 2283…2323 °C |
MDL Number:
MFCD00083151 CAS Number: 12037-01-3 EINECS: 234-856-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...