Gadolinium ≥99.9%, foil

Supplier: STREM CHEMICALS INC MS

93-6435-50X50MM 93-6435-25X25MM
100203-866EA 1292.25 USD
100203-866 100196-802
Gadolinium ≥99.9%, foil
Gadolinium
Formula: Gd
Boiling Pt: 3233 °C
Melting Pt: 1311 °C
Density: 7,898
MDL Number: MFCD00011022
CAS Number: 7440-54-2
EINECS: 231-162-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR