Gadolinium ≥99.9%, ingot

Supplier: STREM CHEMICALS INC MS

93-6432-10G 93-6432-50G
100197-930EA 138.71 USD
100197-930 100200-786
Gadolinium ≥99.9%, ingot
Gadolinium
Formula: Gd
MW: 157.25
Boiling Pt: 3233 °C
Melting Pt: 1311 °C
Density: 7.898
MDL Number: MFCD00011022
CAS Number: 7440-54-2
EINECS: 231-162-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR