Gadolinium ≥99.9%, powder
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-6430-5G
93-6430-25G
100197-928EA
93.42
USD
100197-928
100207-134
Gadolinium ≥99.9%, powder
Gadolinium
Formula:
Gd MW: 157.25 Boiling Pt: 3233 °C Melting Pt: 1311 °C Density: 7.898 |
MDL Number:
MFCD00011022 CAS Number: 7440-54-2 EINECS: 231-162-2 UN: 3089 ADR: 4.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...