Samarium(III) oxide ≥99.99% (Sm basis), PURATREM
Supplier: STREM CHEMICALS INC MS
93-6231-25G
93-6231-5G
100207-300EA
464.25
USD
100207-300
100200-182
Samarium(III) oxide ≥99.99% (Sm basis), PURATREM
Samarium (III) oxide
Formula:
O₃Sm₂ Melting Pt: 2325 °C Density: 8.347 Storage Temperature: Ambient |
MDL Number:
MFCD00011237 CAS Number: 12060-58-1 EINECS: 235-043-6 Merck Index: 14,08347 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...