Praseodymium phosphate
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Praseodymium (III) phosphate, Praseodymium orthophosphate
93-5913-25G
93-5913-5G
100202-814EA
329.78
USD
100202-814
100205-698
Praseodymium phosphate
Praseodymium phosphate
Formula:
O₄PPr |
MDL Number:
MFCD00058789 CAS Number: 14298-31-8 EINECS: 238-231-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...