Lanthanum ≥99.9%, foil
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-5750-50X50MM
93-5750-25X25MM
100208-580EA
900.18
USD
100208-580
100208-198
Lanthanum ≥99.9%, foil
Lanthanum
Formula:
La Boiling Pt: 3454 °C Melting Pt: 920 °C Density: 6.166 Storage Temperature: Ambient |
MDL Number:
MFCD00011066 CAS Number: 7439-91-0 EINECS: 231-099-0 UN: 3208 ADR: 4.3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...