Lanthanum ≥99.9%, powder

Supplier: STREM CHEMICALS INC MS
Danger

93-5745-10G 93-5745-50G
100207-112EA 164.18 USD
100207-112 100196-990
Lanthanum ≥99.9%, powder
Lanthanum
Formula: La
Boiling Pt: 3454 °C
Melting Pt: 885 °C
Density: 6.166
Storage Temperature: Ambient
MDL Number: MFCD00011066
CAS Number: 7439-91-0
EINECS: 231-099-0
UN: 3208
ADR: 4.3,II
Merck Index: 12,05374

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR