Lanthanum(III) oxide ≥99.9% (La basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-5740-250G
93-5740-1000G
100200-592EA
249.11
USD
100200-592
100206-044
Lanthanum(III) oxide ≥99.9% (La basis)
Lanthanum (III) oxide
Formula:
La₂O₃ Boiling Pt: 4200 °C Melting Pt: 2315 °C Density: 6,51 Storage Temperature: Ambient |
MDL Number:
MFCD00011071 CAS Number: 1312-81-8 EINECS: 215-200-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...