Lanthanum(III) nitrate hexahydrate ≥99.9% (La basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-5713-100G
93-5713-500G
100202-212EA
66.52
USD
100202-212
100210-698
Lanthanum(III) nitrate hexahydrate ≥99.9% (La basis)
Lanthanum (III) nitrate hexahydrate
Formula:
La(NO₃)₃·6H₂O Boiling Pt: 126 °C (dec.) Melting Pt: 40 °C Density: 2.347 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149751 CAS Number: 10277-43-7 EINECS: 233-238-0 UN: 1477 ADR: 5.1,II Merck Index: 14,05363 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...