Lanthanum(III) hydroxide, anhydrous ≥99.9% (La basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-5711-50G 93-5711-10G
100199-634EA 181.17 USD
100199-634 100196-768
Lanthanum(III) hydroxide, anhydrous ≥99.9% (La basis)
Lanthanum (III) hydroxide
Formula: H₃LaO₃
Density: 4.28 g/cm³ (20 °C)
MDL Number: MFCD00016160
CAS Number: 14507-19-8
EINECS: 238-510-2
UN: 3262
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR