Barium perchlorate trihydrate ≥99.9% (Ba basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-5650-100G
93-5650-500G
100204-376EA
55.2
USD
100204-376
100211-276
Barium perchlorate trihydrate ≥99.9% (Ba basis)
Barium perchlorate trihydrate
Formula:
BaCl₂O₈·3H₂O Melting Pt: 400 °C (dec.) Density: 2.74 |
MDL Number:
MFCD00149148 CAS Number: 10294-39-0 EINECS: 236-710-4 UN: 1447 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...