Barium titanate ≥99.9% (Ba basis), sintered lumps

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Barium titanate, Barium tantalum oxide, Barium titanate (IV), Barium titanium oxide

93-5639-100G 93-5639-25G
100196-764EA 557.66 USD
100196-764 100197-254
Barium titanate ≥99.9% (Ba basis), sintered lumps
Barium titanate
Formula: BaO₃Ti
Melting Pt: 1620 °C
Density: 5.85
MDL Number: MFCD00003447
CAS Number: 12047-27-7
EINECS: 234-975-0
UN: 1564
ADR: 6.1,III
Merck Index: 12,01029

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR