Barium nitrate ≥99.5%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-5625-500G
93-5625-100G
100208-196EA
144.37
USD
100208-196
100199-142
Barium nitrate ≥99.5%
Barium nitrate
Formula:
Ba(NO₃)₂ Melting Pt: 592 °C Density: 3.230 Storage Temperature: Ambient |
MDL Number:
MFCD00003442 CAS Number: 10022-31-8 EINECS: 233-020-5 UN: 1446 ADR: 5.1,II Merck Index: 13,00986 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...