Barium iodide, anhydrous ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-5619-25G
93-5619-100G
100202-202EA
283.08
USD
100202-202
100197-908
Barium iodide, anhydrous ≥97%
Barium iodide
Formula:
BaI₂ Melting Pt: 714 °C Density: 5.15 g/cm³ (20 °C) |
MDL Number:
MFCD00003452 CAS Number: 13718-50-8 EINECS: 237-276-9 UN: 1564 ADR: 6.1,III Merck Index: 12,01009 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...