Barium hydroxide octahydrate ≥98% ACS
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-5617-1000G
93-5617-250G
100209-446EA
210.89
USD
100209-446
100200-168
Barium hydroxide octahydrate ≥98% ACS
Barium hydroxide octahydrate
Formula:
Ba(OH)₂·8H₂O Boiling Pt: 780 °C (1013 hPa) Melting Pt: 78 °C Density: 2.18 Storage Temperature: Ambient |
MDL Number:
MFCD00149152 CAS Number: 12230-71-6 EINECS: 241-234-5 UN: 2923 ADR: 8,II Merck Index: 13,00980 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...