Tellurium ≥99.999%, powder
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-5220-50G
93-5220-10G
100202-808EA
329.78
USD
100202-808
100205-642
Tellurium ≥99.999%, powder
Tellurium
Formula:
Te Boiling Pt: 989.8 °C Melting Pt: 449.5 °C Density: 6.24 |
MDL Number:
MFCD00134062 CAS Number: 13494-80-9 EINECS: 236-813-4 UN: 3288 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...