Indium ≥99.9%, shot

Supplier: STREM CHEMICALS INC MS
Warning

93-4942-25G 93-4942-5G
100201-120EA 189.66 USD
100201-120 100199-124
Indium ≥99.9%, shot
Indium
Formula: In
MW: 114.82 g/mol
Boiling Pt: 2000
Melting Pt: 156
Density: 7.31
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR