Silver(I) telluride ≥99.99% (Ag/Te basis), PURATREM
Supplier: STREM CHEMICALS INC MS
Synonyms:
Silver telluride
93-4733-1G
93-4733-5G
100204-336EA
69.35
USD
100204-336
100210-762
Silver(I) telluride ≥99.99% (Ag/Te basis), PURATREM
Silver (I) telluride
Formula:
Ag₂Te Melting Pt: 955 °C Density: 8.5 |
MDL Number:
MFCD00014146 CAS Number: 12002-99-2 EINECS: 234-419-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...